Learn More
Biomatik Corporation Rat Erythropoietin Receptor (EPOR) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03968
Encompass_Preferred
Rat Erythropoietin Receptor (EPOR) ELISA Kit for detecting EPOR in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 62.5-4,000pg/ml. Sensitivity: 26.9pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q07303. Target Synonyms: EPO-R. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2423