Learn More
Biomatik Corporation Escherichia coli Protein (ECP) ELISA Kit, 96 tests
Catalog No. 501502424
Encompass_Preferred
The Escherichia coli Protein (ECP) ELISA Kit allows for quantitative determination of Escherichia coli Protein in Biological agents. Species Reactivity: All species. Detection Range: 0.312-20 ng/mL. Sensitivity: 0.125 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P23827. Cross Activity: No significant cross-reactivity or interference between Escherichia coli Protein and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2424 Supplier Biomatik Corporation Supplier No. EKU03969
The Escherichia coli Protein (ECP) ELISA Kit allows for quantitative determination of Escherichia coli Protein in Biological agents. Species Reactivity: All species. Detection Range: 0.312-20 ng/mL. Sensitivity: 0.125 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P23827. Cross Activity: No significant cross-reactivity or interference between Escherichia coli Protein and analogues was observed. Restriction: For Research Use Only.