Learn More
Biomatik Corporation Estradiol (E2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03971
Encompass_Preferred
The Estradiol (E2) ELISA Kit allows for quantitative determination of Estradiol in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: All species. Detection Range: 12.35-1000 pg/mL. Sensitivity: 4.45 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Competitive Inhibition. Detection Method: Colorimetric. Assay Time: 2 hours. Cross Activity: No significant cross-reactivity or interference between Estradiol and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2425