Learn More
Biomatik Corporation Human Estrogen Receptor Alpha (ERa) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03973
Encompass_Preferred
The Human Estrogen Receptor Alpha (ERa) ELISA Kit allows for quantitative determination of Estrogen Receptor Alpha in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.239 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P03372. Cross Activity: No significant cross-reactivity or interference between Estrogen Receptor Alpha and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2426