Learn More
Biomatik Corporation Mouse Estrogen Receptor Alpha (ERa) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03974
Encompass_Preferred
Mouse Estrogen Receptor Alpha (ERa) ELISA Kit for detecting ERa in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.122ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19785. Gene ID: 13982. Target Synonyms: ER-A1; ER; ESR; ESR1; ESRA; NR3A1; NR3-A1; Estrogen Receptor 1; Nuclear Receptor Subfamily 3,Group A,Member 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2427