Learn More
Biomatik Corporation Human Defensin Beta 103B (DEFB103B) ELISA Kit
Supplier:  Biomatik Corporation EKU03662
Encompass_Preferred
Human Defensin Beta 103B (DEFB103B) ELISA Kit for detecting DEFB103B in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2,000pg/ml. Sensitivity: 12.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P81534. Gene ID: 414325. Target Synonyms: HBP3; HBD3; BD3; DEFB103; DEFB3; Defensin-like protein; Beta-defensin 3. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5955