Learn More
Biomatik Corporation Human Estrogen Receptor Beta (ERb) ELISA Kit, 96 tests
Catalog No. 501502428
Encompass_Preferred
The Human Estrogen Receptor Beta (ERb) ELISA Kit allows for quantitative determination of Estrogen Receptor Beta in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.127 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q92731. Cross Activity: No significant cross-reactivity or interference between Estrogen Receptor Beta and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2428 Supplier Biomatik Corporation Supplier No. EKU03976
The Human Estrogen Receptor Beta (ERb) ELISA Kit allows for quantitative determination of Estrogen Receptor Beta in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.127 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q92731. Cross Activity: No significant cross-reactivity or interference between Estrogen Receptor Beta and analogues was observed. Restriction: For Research Use Only.