Learn More
Biomatik Corporation Human Early Growth Response Protein 1 (EGR1) ELISA Kit
Supplier:  Biomatik Corporation EKU03825
Encompass_Preferred
Human Early Growth Response Protein 1 (EGR1) ELISA Kit for detecting EGR1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P18146. Gene ID: 1958. Target Synonyms: Zif268; AT225; G0S30; KROX24; NGFI-A; TIS8; ZIF-268; ZNF225; NGFIA; TIS8; ZENK; Nerve Growth Factor-Induced Protein A; Transcription Factor ETR103; Zinc Finger Protein 225. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5969