Learn More
Biomatik Corporation Mouse Excitatory Amino Acid Transporter 2 (EAAT2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU03992
Encompass_Preferred
Mouse Excitatory Amino Acid Transporter 2 (EAAT2) ELISA Kit for detecting EAAT2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.065ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P43006. Gene ID: 20511. Target Synonyms: SLC1A2; EAAT2; GLT-1; Solute Carrier Family 1 Member 2; Glutamate/aspartate transporter II; Sodium-dependent glutamate/aspartate transporter 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2431