Learn More
Biomatik Corporation Rat Elastase 2A (ELA2A) ELISA Kit
Supplier:  Biomatik Corporation EKU03845
Encompass_Preferred
Rat Elastase 2A (ELA2A) ELISA Kit for detecting ELA2A in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.119ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P00774. Gene ID: 24332. Target Synonyms: ELA1; PE-1; CELA2A; Chymotrypsin-Like Elastase Family Member 2A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5974