Learn More
Biomatik Corporation Human Extracellular Matrix Metalloproteinase Inducer (EMMPRIN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04002
Encompass_Preferred
The Human Extracellular Matrix Metalloproteinase Inducer (EMMPRIN) ELISA Kit allows for quantitative determination of Extracellular Matrix Metalloproteinase Inducer in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.2-2000 pg/mL. Sensitivity: 11.8 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P35613. Cross Activity: No significant cross-reactivity or interference between Extracellular Matrix Metalloproteinase Inducer and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2433