Learn More
Biomatik Corporation Human Endostatin (ES) ELISA Kit
Supplier:  Biomatik Corporation EKU03878
Encompass_Preferred
Human Endostatin (ES) ELISA Kit for detecting ES in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-40ng/ml. Sensitivity: 0.255ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P39060. Gene ID: 80781. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5978