Learn More
Biomatik Corporation Epitestosterone (ET) ELISA Kit
Supplier:  Biomatik Corporation EKU03948
Encompass_Preferred
Epitestosterone (ET) ELISA Kit for detecting ET in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 39.06-10,000pg/ml. Sensitivity: 15.29pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5987