Learn More
Biomatik Corporation Human Family With Sequence Similarity 132, Member A (FAM132A) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04015
Encompass_Preferred
The Human Family With Sequence Similarity 132, Member A (FAM132A) ELISA Kit allows for quantitative determination of Family With Sequence Similarity 132, Member A in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.054 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q5T7M4. Cross Activity: No significant cross-reactivity or interference between Family With Sequence Similarity 132, Member A and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2437