Learn More
Biomatik Corporation Mouse Estrogen Receptor Beta (ERb) ELISA Kit
Supplier:  Biomatik Corporation EKU03977
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.312-20ng/mL. Sensitivity: 0.115ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: O08537. Target Synonyms: ER-B; ESRB; ESTRB; NR3A2; NR3-A2; ESR2; Estrogen Receptor 2; Nuclear Receptor Subfamily 3,Group A,Member 2. Restriction: For Research Use Only.
Catalog No. 50-228-5989