Learn More
Biomatik Corporation Human Family With Sequence Similarity 3, Member B (FAM3B) ELISA Kit, 96 tests
Catalog No. 501502438
Encompass_Preferred
The Human Family With Sequence Similarity 3, Member B (FAM3B) ELISA Kit allows for quantitative determination of Family With Sequence Similarity 3, Member B in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.255 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P58499. Cross Activity: No significant cross-reactivity or interference between Family With Sequence Similarity 3, Member B and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2438 Supplier Biomatik Corporation Supplier No. EKU04018
The Human Family With Sequence Similarity 3, Member B (FAM3B) ELISA Kit allows for quantitative determination of Family With Sequence Similarity 3, Member B in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.255 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P58499. Cross Activity: No significant cross-reactivity or interference between Family With Sequence Similarity 3, Member B and analogues was observed. Restriction: For Research Use Only.