Learn More
Biomatik Corporation Ethinylestradiol (EE) ELISA Kit
Supplier:  Biomatik Corporation EKU03983
Encompass_Preferred
Ethinylestradiol (EE) ELISA Kit for detecting EE in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 24.69-2,000pg/ml. Sensitivity: 9.13pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Target Synonyms: Ethinyl Estradiol ; Ethynyl Estradiol; Ethinyl Oestradiol. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5991