Learn More
Biomatik Corporation Pig Fatty Acid Binding Protein 1, Liver (FABP1) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04029
Encompass_Preferred
Pig Fatty Acid Binding Protein 1 (FABP1) ELISA Kit for detecting FABP1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49924. Gene ID: 445535. Target Synonyms: FABP-1; FABPL; L-FABP; LFABP; Liver-type fatty acid-binding protein; Fatty Acid Binding Protein 1, Liver. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2440