Learn More
Biomatik Corporation Human Fascin (FSCN) ELISA Kit
Supplier:  Biomatik Corporation EKU04027
Encompass_Preferred
Human Fascin (FSCN) ELISA Kit for detecting FSCN in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.113ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q16658. Gene ID: 6624. Target Synonyms: FSCN1; SNL; p55; Singed-like protein; 55 kDa actin-bundling protein; Fascin homolog 1, Actin-Bundling Protein (Strongylocentrotus Purpuratus) ; Singed,Drosophila,Homolog-Like. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5995