Learn More
Biomatik Corporation Pig Ferritin (FE) ELISA Kit
Supplier:  Biomatik Corporation EKU04067
Encompass_Preferred
Pig Ferritin (FE) ELISA Kit for detecting FE in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 0.94-60ng/ml. Sensitivity: 0.39ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-5997