Learn More
Biomatik Corporation Rat Fatty Acid Binding Protein 2, Intestinal (FABP2) ELISA Kit, 96 tests
Catalog No. 501502443
Encompass_Preferred
Rat Fatty Acid Binding Protein 2, Intestinal (FABP2) ELISA Kit for detecting FABP2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.133ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02693. Gene ID: 25598. Target Synonyms: FABPI; IFABP; I-FABP; Intestinal-type fatty acid-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2443 Supplier Biomatik Corporation Supplier No. EKU04033
Rat Fatty Acid Binding Protein 2, Intestinal (FABP2) ELISA Kit for detecting FABP2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.133ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P02693. Gene ID: 25598. Target Synonyms: FABPI; IFABP; I-FABP; Intestinal-type fatty acid-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.