Learn More
Biomatik Corporation Human Fatty Acid Binding Protein 3, Muscle And Heart (FABP3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04034
Encompass_Preferred
The Human Fatty Acid Binding Protein 3, Muscle And Heart (FABP3) ELISA Kit allows for quantitative determination of Fatty Acid Binding Protein 3, Muscle And Heart in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.625-40 ng/mL. Sensitivity: 0.251 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P05413. Cross Activity: No significant cross-reactivity or interference between Fatty Acid Binding Protein 3, Muscle And Heart and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2444