Learn More
Biomatik Corporation Human Fibrinopeptide A (FPA) ELISA Kit
Supplier:  Biomatik Corporation EKU04112
Encompass_Preferred
Human Fibrinopeptide A (FPA) ELISA Kit for detecting FPA in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.23-100ng/ml. Sensitivity: 0.55ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P02671. Gene ID: 2243. Target Synonyms: FP-A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6005