Learn More
Biomatik Corporation Rat Fatty Acid Binding Protein 3, Muscle And Heart (FABP3) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04037
Encompass_Preferred
Rat Heart-type Fatty Acid Binding Protein (H-FABP) ELISA Kit for detecting H-FABP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.78-50ng/ml. Sensitivity: 0.32ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07483. Gene ID: 79131. Target Synonyms: MDGI; FABP11; H-FABP; O-FABP; Hfabp; FABP3; Fatty Acid Binding Protein 3, Muscle And Heart; Mammary Derived Growth Inhibitor; Fatty Acid Binding Protein 11; Muscle fatty acid-binding protein; Heart-ty. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2446