Learn More
Biomatik Corporation Human Fibroblast Activation Protein Alpha (FAPa) ELISA Kit
Supplier:  Biomatik Corporation EKU04117
Encompass_Preferred
Human Fibroblast Activation Protein Alpha (FAPa) ELISA Kit for detecting FAPa in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.78-50ng/ml. Sensitivity: 0.29ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q12884. Gene ID: 2191. Target Synonyms: APCE; SIMP; DPPIV; Seprase; 170 kDa melanoma membrane-bound gelatinase; Post-proline cleaving enzyme; Serine integral membrane protease; Surface-expressed protease. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6007