Learn More
Biomatik Corporation Human Fatty Acid Binding Protein 4, Adipocyte (FABP4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04038
Encompass_Preferred
The Human Fatty Acid Binding Protein 4, Adipocyte (FABP4) ELISA Kit allows for quantitative determination of Fatty Acid Binding Protein 4, Adipocyte in Serum, Plasma, Tissue homogenates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 31.25-2000 pg/mL. Sensitivity: 12.6 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P15090. Cross Activity: No significant cross-reactivity or interference between Fatty Acid Binding Protein 4, Adipocyte and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2447