Learn More
Biomatik Corporation Human Fibromodulin (FMOD) ELISA Kit
Supplier:  Biomatik Corporation EKU04196
Encompass_Preferred
Human Fibromodulin (FMOD) ELISA Kit for detecting FMOD in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.2-2,000pg/ml. Sensitivity: 13.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q06828. Gene ID: 2331. Target Synonyms: SLRR2E; FM; Collagen-binding 59 kDa protein; Keratan sulfate proteoglycan fibromodulin; KSPG fibromodulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6011