Learn More
Biomatik Corporation Mouse Fatty Acid Binding Protein 4, Adipocyte (FABP4) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04039
Encompass_Preferred
Mouse Fatty Acid Binding Protein 4 (FABP4) ELISA Kit for detecting FABP4 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P04117. Gene ID: 11770. Target Synonyms: A-FABP; AFABP; AP2; Fatty Acid Binding Protein 4, Adipocyte; Adipocyte Protein 2; Adipocyte-type fatty acid-binding protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2448