Learn More
Biomatik Corporation Rat Focal Adhesion Kinase (FAK) ELISA Kit
Supplier:  Biomatik Corporation EKU04243
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates and other biological fluids. Species Reactivity: Rat. Detection Range: 0.312-20ng/mL. Sensitivity: 0.119ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: Not Available. Target Synonyms: PTK2; FADK; FAK1; Pp125FAK; PPP1R71; Protein Tyrosine Kinase 2; Focal adhesion kinase-related nonkinase; Protein phosphatase 1 regulatory subunit 71. Restriction: For Research Use Only.
Catalog No. 50-228-6016