Learn More
Biomatik Corporation Human Fatty Acid Binding Protein 5, Epidermal (FABP5) ELISA Kit, 96 tests
Catalog No. 501502450
Encompass_Preferred
The Human Fatty Acid Binding Protein 5, Epidermal (FABP5) ELISA Kit allows for quantitative determination of Fatty Acid Binding Protein 5, Epidermal in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.115 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q01469. Cross Activity: No significant cross-reactivity or interference between Fatty Acid Binding Protein 5, Epidermal and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2450 Supplier Biomatik Corporation Supplier No. EKU04041
The Human Fatty Acid Binding Protein 5, Epidermal (FABP5) ELISA Kit allows for quantitative determination of Fatty Acid Binding Protein 5, Epidermal in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.115 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: Q01469. Cross Activity: No significant cross-reactivity or interference between Fatty Acid Binding Protein 5, Epidermal and analogues was observed. Restriction: For Research Use Only.