Learn More
Biomatik Corporation Free Triiodothyronine (fT3) ELISA Kit
Supplier:  Biomatik Corporation EKU04275
Encompass_Preferred
Free Triiodothyronine (fT3) ELISA Kit for detecting fT3 in serum, plasma, other biological fluids. Species Reactivity: Universal. Detection Range: 740.7-60000fg/ml. Sensitivity: 249.5fg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6021