Learn More
Biomatik Corporation Human Fatty Acid Binding Protein 7, Brain (FABP7) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04045
Encompass_Preferred
The Human Fatty Acid Binding Protein 7, Brain (FABP7) ELISA Kit allows for quantitative determination of Fatty Acid Binding Protein 7, Brain in Serum, Plasma and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.47-30 ng/mL. Sensitivity: 0.18 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O15540. Cross Activity: No significant cross-reactivity or interference between Fatty Acid Binding Protein 7, Brain and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2453