Learn More
Biomatik Corporation Rat Galanin (GAL) ELISA Kit
Supplier:  Biomatik Corporation EKU04312
Encompass_Preferred
Rat Galanin (GAL) ELISA Kit for detecting GAL in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 12.35-1,000pg/ml. Sensitivity: 5.34pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P10683. Gene ID: 29141. Target Synonyms: GALN; GLNN; GMAP; Galanin Prepropeptide; Galanin-Message-Associated Peptide. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6026