Learn More
Biomatik Corporation Human Gastrin Releasing Peptide (GRP) ELISA Kit
Supplier:  Biomatik Corporation EKU04349
Encompass_Preferred
Human Gastrin Releasing Peptide (GRP) ELISA Kit for detecting GRP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 12.35-1,000pg/ml. Sensitivity: 4.99pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P09758. Gene ID: 4070. Target Synonyms: BN; GRP10; Bombesin; Neuromedin C. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6028