Learn More
Biomatik Corporation Rat Fatty Acid Desaturase 2 (FADS2) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04050
Encompass_Preferred
Rat Fatty Acid Desaturase 2 (FADS2) ELISA Kit for detecting FADS2 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 1.56-100ng/ml. Sensitivity: 0.64ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9Z122. Gene ID: 83512. Target Synonyms: LLCDL2; FADSD6; D6D; TU13; DES6; Delta-6-Desaturase; Delta (6) fatty acid desaturase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2455