Learn More
Biomatik Corporation Human Glucose-6-phosphate Dehydrogenase (G6PD) ELISA Kit
Supplier:  Biomatik Corporation EKU04406
Encompass_Preferred
Human Glucose-6-phosphate Dehydrogenase (G6PD) ELISA Kit for detecting G6PD in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 3.12-200ng/ml. Sensitivity: 1.18ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11413. Gene ID: 2539. Target Synonyms: Glucose-6-phosphate 1-dehydrogenase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6034