Learn More
Biomatik Corporation Mouse Fatty Acid Synthase (FASN) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04054
Encompass_Preferred
Mouse Fatty Acid Synthase (FASN) ELISA Kit for detecting FASN in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19096. Gene ID: 14104. Target Synonyms: FAS; OA519; SDR27X1; Short Chain Dehydrogenase/Reductase Family 27X,Member 1; S-acetyltransferase; 3-oxoacyl synthase; Oleoyl hydrolase; Enoyl-acyl-carrier-protein reductase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2457