Learn More
Biomatik Corporation Mouse Glucose Transporter 4 (GLUT4) ELISA Kit
Supplier:  Biomatik Corporation EKU04420
Encompass_Preferred
Sample Type: Tissue homogenates, cell lysates, cell culture supernates and other biological fluids. Species Reactivity: Mouse. Detection Range: 0.156-10ng/mL. Sensitivity: 0.063ng/mL. Format: 96-Well Plate. Assay Type: Sandwich. Detection Method: Sandwich. Assay Time: 3 hours. UniProt ID: P14142. Target Synonyms: SLC2A4; Solute carrier family 2, facilitated glucose transporter member 4; Glucose transporter type 4, insulin-responsive. Restriction: For Research Use Only.
Catalog No. 50-228-6036