Learn More
Biomatik Corporation Rat Fatty Acid Synthase (FASN) ELISA Kit, 96 tests
Catalog No. 501502458
Encompass_Preferred
Rat Fatty Acid Synthase (FASN) ELISA Kit for detecting FASN in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P12785. Gene ID: 50671. Target Synonyms: FAS; OA519; SDR27X1; Short Chain Dehydrogenase/Reductase Family 27X,Member 1; S-acetyltransferase; 3-oxoacyl synthase; Oleoyl hydrolase; Enoyl-acyl-carrier-protein reductase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2458 Supplier Biomatik Corporation Supplier No. EKU04055
Add to Cart
Add to Cart
Rat Fatty Acid Synthase (FASN) ELISA Kit for detecting FASN in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 0.312-20ng/ml. Sensitivity: 0.125ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P12785. Gene ID: 50671. Target Synonyms: FAS; OA519; SDR27X1; Short Chain Dehydrogenase/Reductase Family 27X,Member 1; S-acetyltransferase; 3-oxoacyl synthase; Oleoyl hydrolase; Enoyl-acyl-carrier-protein reductase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.