Learn More
Biomatik Corporation Human Glutaminase (GLS) ELISA Kit
Supplier:  Biomatik Corporation EKU04461
Encompass_Preferred
Human Glutaminase (GLS) ELISA Kit for detecting GLS in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.119ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O94925. Gene ID: 2744. Target Synonyms: GLS1; Glutaminase kidney isoform, mitochondrial; K-glutaminase; L-glutamine amidohydrolase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6039