Learn More
Biomatik Corporation Human Glutathione S Transferase A4 (GSTA4) ELISA Kit
Supplier:  Biomatik Corporation EKU04491
Encompass_Preferred
Human Glutathione S Transferase A4 (GSTA4) ELISA Kit for detecting GSTA4 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: O15217. Gene ID: 2941. Target Synonyms: GST-A4; GSTA4-4; GTA4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6043