Learn More
Biomatik Corporation Human Fc Fragment of IgG Low Affinity IIIa Receptor (FcgR3A) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04063
Encompass_Preferred
The Human Fc Fragment of IgG Low Affinity IIIa Receptor (FcgR3A) ELISA Kit allows for quantitative determination of Fc Fragment of IgG Low Affinity IIIa Receptor in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 125-2000 ng/mL. Sensitivity: 50 pg/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P08637. Cross Activity: No significant cross-reactivity or interference between Fc Fragment of IgG Low Affinity IIIa Receptor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2461