Learn More
Biomatik Corporation Human Fc Fragment of IgG Low Affinity IIIb Receptor (FcgR3B) ELISA Kit, 96 tests
Catalog No. 501502462
Encompass_Preferred
The Human Fc Fragment of IgG Low Affinity IIIb Receptor (FcgR3B) ELISA Kit allows for quantitative determination of Fc Fragment of IgG Low Affinity IIIb Receptor in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.135 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O75015. Cross Activity: No significant cross-reactivity or interference between Fc Fragment of IgG Low Affinity IIIb Receptor and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2462 Supplier Biomatik Corporation Supplier No. EKU04064
The Human Fc Fragment of IgG Low Affinity IIIb Receptor (FcgR3B) ELISA Kit allows for quantitative determination of Fc Fragment of IgG Low Affinity IIIb Receptor in Serum, Plasma, Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.135 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: O75015. Cross Activity: No significant cross-reactivity or interference between Fc Fragment of IgG Low Affinity IIIb Receptor and analogues was observed. Restriction: For Research Use Only.