Learn More
Biomatik Corporation Mouse Heat Shock 70kDa Protein 5 (HSPA5) ELISA Kit
Supplier:  Biomatik Corporation EKU04661
Encompass_Preferred
Mouse Heat Shock 70kDa Protein 5 (HSPA5) ELISA Kit for detecting HSPA5 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Mouse (Mus musculus). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P20029. Gene ID: 14828. Target Synonyms: MIF2; BIP; GRP78; Immunoglobulin heavy chain-binding protein; Glucose Regulated Protein 78; Binding Immunoglobulin Protein; Endoplasmic reticulum lumenal Ca binding grp78. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6061