Learn More
Biomatik Corporation Human Fc Fragment of IgG Receptor Transporter Alpha (FcgRT) ELISA Kit, 96 tests
Catalog No. 501502463
Encompass_Preferred
The Human Fc Fragment of IgG Receptor Transporter Alpha (FcgRT) ELISA Kit allows for quantitative determination of Fc Fragment of IgG Receptor Transporter Alpha in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.057 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P55899. Cross Activity: No significant cross-reactivity or interference between Fc Fragment of IgG Receptor Transporter Alpha and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2463 Supplier Biomatik Corporation Supplier No. EKU04065
The Human Fc Fragment of IgG Receptor Transporter Alpha (FcgRT) ELISA Kit allows for quantitative determination of Fc Fragment of IgG Receptor Transporter Alpha in Tissue homogenates, Cell lysates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.057 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P55899. Cross Activity: No significant cross-reactivity or interference between Fc Fragment of IgG Receptor Transporter Alpha and analogues was observed. Restriction: For Research Use Only.