Learn More
Biomatik Corporation Human Ferritin (FE) ELISA Kit, 96 tests
Catalog No. 501502464
Encompass_Preferred
The Human Ferritin (FE) ELISA Kit allows for quantitative determination of Ferritin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.061 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02794. Cross Activity: No significant cross-reactivity or interference between Ferritin and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2464 Supplier Biomatik Corporation Supplier No. EKU04066
The Human Ferritin (FE) ELISA Kit allows for quantitative determination of Ferritin in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.156-10 ng/mL. Sensitivity: 0.061 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02794. Cross Activity: No significant cross-reactivity or interference between Ferritin and analogues was observed. Restriction: For Research Use Only.