Learn More
Biomatik Corporation Rat Heparin Binding Epidermal Growth Factor Like Growth Factor (HBEGF) ELISA Kit
Supplier:  Biomatik Corporation EKU04742
Encompass_Preferred
Rat Heparin Binding Epidermal Growth Factor Like Growth Factor (HBEGF) ELISA Kit for detecting HBEGF in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q06175. Gene ID: 25433. Target Synonyms: HB-EGF; DTR; DTS; DTSF; HEGFL; Diphtheria Toxin Receptor; Proheparin-binding EGF-like growth factor; Diphtheria toxin receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6066