Learn More
Biomatik Corporation Rat Ferritin, Heavy Polypeptide (FTH) ELISA Kit, 96 tests
Supplier:  Biomatik Corporation EKU04071
Encompass_Preferred
Rat Ferritin, Heavy Polypeptide (FTH) ELISA Kit for detecting FTH in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Rat (Rattus norvegicus). Detection Range: 3.12-200ng/ml. Sensitivity: 1.15ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P19132. Gene ID: 25319. Target Synonyms: FTH1; FTHL6; PIG15; PLIF; Ferritin Heavy Chain; Apoferritin; Placenta Immunoregulatory Factor; Proliferation-Inducing Protein 15; Cell proliferation-inducing gene 15 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-150-2467