Learn More
Biomatik Corporation Pig High Mobility Group Protein 1 (HMGB1) ELISA Kit
Supplier:  Biomatik Corporation EKU04791
Encompass_Preferred
Pig High Mobility Group Protein 1 (HMGB1) ELISA Kit for detecting HMGB1 in serum, plasma, other biological fluids. Species Reactivity: Pig (Sus scrofa; Porcine). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.1pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: F2Z594. Target Synonyms: HMG1; HMG3; SBP1; Sulfoglucuronyl Carbohydrate Binding Protein; Amphoterin; High Mobility Group Box 1 Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-228-6074