Learn More
Biomatik Corporation Human Ferritin, Light Polypeptide (FTL) ELISA Kit, 96 tests
Catalog No. 501502468
Encompass_Preferred
The Human Ferritin, Light Polypeptide (FTL) ELISA Kit allows for quantitative determination of Ferritin, Light Polypeptide in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.129 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02792. Cross Activity: No significant cross-reactivity or interference between Ferritin, Light Polypeptide and analogues was observed. Restriction: For Research Use Only.
Catalog No. 50-150-2468 Supplier Biomatik Corporation Supplier No. EKU04072
The Human Ferritin, Light Polypeptide (FTL) ELISA Kit allows for quantitative determination of Ferritin, Light Polypeptide in Serum, Plasma, Tissue homogenates, Cell lysates, Cell culture supernates and other biological fluids. Species Reactivity: Homo sapiens (Human). Detection Range: 0.312-20 ng/mL. Sensitivity: 0.129 ng/mL. Format: 96-Well Plate. Assay Type: ELISA, Double-antibody Sandwich. Detection Method: Colorimetric. Assay Time: 3 hours. UniProt ID: P02792. Cross Activity: No significant cross-reactivity or interference between Ferritin, Light Polypeptide and analogues was observed. Restriction: For Research Use Only.